Learn More
Aviva Systems Biology Corp SCAMP1 Antibody - middle region (ARP59465_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59465_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCAMP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SCAMP1. Alternate names: SCAMP, SCAMP37ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-705