Learn More
Aviva Systems Biology Corp SCAMP3 Antibody - N-terminal region (ARP59468_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59468_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCAMP3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SCAMP3. Alternate names: C1orf3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-707