Learn More
Aviva Systems Biology Corp Scamp5 Antibody - C-terminal region (ARP59469_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59469_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Scamp5. It was validated on Western Blot. . Alternate names: AI426171, AW558254, Sc5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-708