Learn More
Interscience Scan 1200 Adapter
SDP
Catalog No. 16948931 Shop All Interscience Products
Change view
Click to view available options
Holds:
Compact Dry
MC-Media Pad
Petrifilm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
16-948-931 Petrifilm
16-948-929 MC-Media Pad
16-948-928 Compact Dry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-948-931 Supplier Interscience Supplier No. 437002
Add to Cart
Add to Cart
Adapters for Scan 1200 automatic colony counter.
  • Adapter for use with Scan 1200
  • Available for MC-Media Pad™, Petrifilm™, or Compact Dry™
Specifications
Product Type Dish Centering Adapter
Display Type None
Holds Petrifilm
For Use With (Equipment) Scan 1200