Learn More
Interscience Scan 300/500/1200 Adapter
SDP
Catalog No. 16948930 Shop All Interscience Products
Change view
Click to view available options
Display Type:
None
Catalog No. Display Type
16-948-930 None
1 options
Catalog No. 16-948-930 Supplier Interscience Supplier No. 436005
Add to Cart
Add to Cart
Adapter for 55 mm Petri dish for Scan 300, Scan 500, and Scan 1200 automated colony counters.
  • Adapter for 55 mm dia. Petri dishes
  • For use with Scan 300, Scan 500, or Scan 1200
Specifications
Product Type Dish Centering Adapter
Display Type None
Holds 55 mm dia. Petri Dish
For Use With (Equipment) Scan 300/500/1200