Learn More
Interscience Scan 300
SDP
Catalog No. 16949090 Shop All Interscience Products
Change view
Click to view available options
Display Type:
LED
Catalog No. Display Type
16-949-090 LED
1 options
Catalog No. 16-949-090 Supplier Interscience Supplier No. 436300
Add to Cart
Add to Cart
Request a Quote
The Scan 300 is an automatic colony counter with basic features.
Scan 300 is an automatic colony counter that includes the essential features for Petri dish counting. It is specifically adapted to the usual media in food microbiology: PCA, MRS, etc.
  • Minimum size of detected colony: 0.1 mm
  • Camera: 1 megapixel to zoom x 28
  • Counting on round Petri dishes of 55 to 90 dia. mm
  • Counting on pour, surface, Spiral, circle mode plated dishes
  • Language: English, French, Japanese, Chinese, Russian, Spanish, German
Specifications
Product Type Automated Colony Counter
Display Type LED
Voltage 100/240 V