Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience Scan 50 Counting Grids
SDP

Catalog No. 16948960 Shop All Interscience Products
Change view
Click to view available options
Product Line:
Easy Spiral
EasySpiral Dilute
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line
16-948-960 EasySpiral Dilute
16-948-958 Easy Spiral
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-948-960 Supplier Interscience Supplier No. 435038
Add to Cart
Add to Cart

Counting grids for use with Scan 50 and Scan 50 Pro.

  • EasySpiral and EasySpiral Dilute circle counting grids
  • Diameter: 90 mm
  • 1 transparent and 1 white diffusing background for Scan 50 and Scan 50 Pro

Specifications

Product Type Counting Grids
Display Type None
Color Transparent and White
Product Line EasySpiral Dilute
Quantity 2/Pk.
Diameter (Metric) 90 mm
For Use With (Equipment) Scan 50
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.