Learn More
Interscience Scan 5000 Ai
SDP
Catalog No. 01670881 Shop All Interscience Products
Change view
Click to view available options
Display Type:
LED
Catalog No. Display Type
01-670-881 LED
1 options
A perpetual software license (01-670-884) is required for operation and must be purchased with this unit.
Catalog No. 01-670-881 Supplier Interscience Supplier No. 434500
Add to Cart
Add to Cart
Request a Quote
A perpetual software license (01-670-884) is required for operation and must be purchased with this unit.
Automatic AI colony counter and inhibition zone reader for up to 150 mm plates
  • Scan 5000 Ai automates and standardizes colony counting for microbiological analyses.
  • It offers strong performance and continuous development opportunities at the cutting edge of technological developments.
  • It is suitable for all sizes of Petri dishes up to 150 mm and all culture media.
Specifications
Product Type Automated Colony Counter and Zone Measurement System
Display Type LED
Voltage 100/240 V