Learn More
MEDTOX™ MiniPet Pipettor PV
Catalog No. 22000407
Change view
Click to view available options
Type:
Precision Pipette
Catalog No. Type
22-000-407 Precision Pipette
1 options
This item is not returnable. View return policy
Catalog No. 22-000-407 Supplier MEDTOX™ Supplier No. 833077
Add to Cart
Add to Cart
This item is not returnable. View return policy
Precision pipette used with pipette tips to dispense urine on to PROFILE-V devices
  • Designed for use with PROFILE™-V MEDTOX Scan Test system
  • Dispenses 75μL accurately when using the appropriate technique
  • Store MiniPet in a dry, secure location, at room temperature (18 - 25°C or 64 - 77°F)
Specifications
Type Precision Pipette
For Use With (Application) Dispense urine on to PROFILE-V devices
Quantity Each