Learn More
Aviva Systems Biology Corp SCAND1 Antibody - N-terminal region (ARP37772_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37772_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCAND1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SCAND1. Alternate names: RAZ1, SDP1ApplicationWB. Species reactivity: Horse, Human, Pig
Catalog No. 50-014-56429