Learn More
Aviva Systems Biology Corp SCAND2 Antibody - N-terminal region (ARP36017_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36017_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCAND2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human SCAND2. Alternate names: SCAND2, ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55971