Learn More
Aviva Systems Biology Corp ZBED9 Antibody - C-terminal region (ARP47706_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47706_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCAND3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SCAND3. Alternate names: DKFZp434N092, FLJ31087, KIAA1925, MGC131782, MGC133310, MGC133311, SCAND3, ZFP38-L, ZNF305P2, dJ1186N24.3, ZNF452ApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-59646