Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scandium Standard For ICP, 1000μg/mL, SPEX CertiPrep™
SDP

Catalog No. PLSC22T Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PLSC22T 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PLSC22T Supplier SPEX CertiPrep Supplier No. PLSC22T
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sc1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 12060-08-1 , 7697-37-2 , 7732-18-5
Molecular Formula O3Sc2
Molecular Weight (g/mol) 137.91
MDL Number MFCD00011223
InChI Key HJGMWXTVGKLUAQ-UHFFFAOYSA-N
IUPAC Name discandium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sc+3].[Sc+3]

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 2.0, 97.7, 0.3
Analyte or Component Names Sc
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Sc
Solvent or Matrix 2% HNO3
Quantity 250 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material Scandium Standard, For AA and ICP
Show More Show Less
Hazard Category Causes severe skin burns and eye damage May cause respiratory irritation
DOTInformation Hazardous
Recommended Storage Ambient
WARNING: Cancer - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.