Learn More
Strem, An Ascensus Company CAS# 20662-14-0. 1g. Scandium(III) chloride hexahydrate (99.9%-Sc) (REO). MFCD00149852
SDP
Catalog No. 5090114402
Encompass
CAS# 20662-14-0. 1g. Scandium(III) chloride hexahydrate (99.9%-Sc) (REO). MFCD00149852. Molecular Weight: 151.32 (259.41). Molecular Formula: ScCl3∙6H2O. Color/form: white xtl. Strem# 93-2111. http://www.strem.com/catalog/v/93-2111/
Catalog No. 50-901-14402 Supplier Strem, An Ascensus Company Supplier No. 9321111G
CAS# 20662-14-0. 1g. Scandium(III) chloride hexahydrate (99.9%-Sc) (REO). MFCD00149852. Molecular Weight: 151.32 (259.41). Molecular Formula: ScCl3∙6H2O. Color/form: white xtl. Strem# 93-2111. http://www.strem.com/catalog/v/93-2111/