Learn More
Strem, An Ascensus Company CAS# 13465-60-6. 1g. Scandium(III) nitrate pentahydrate (99.9%-Sc) (REO). MFCD00149849
SDP
Catalog No. 5090114396
Encompass
CAS# 13465-60-6. 1g. Scandium(III) nitrate pentahydrate (99.9%-Sc) (REO). MFCD00149849. Molecular Weight: 230.97 (321.05). Molecular Formula: Sc(NO3)3∙5H2O. Color/form: white xtl. Strem# 93-2104. http://www.strem.com/catalog/v/93-2104/
Catalog No. 50-901-14396 Supplier Strem, An Ascensus Company Supplier No. 9321041G
CAS# 13465-60-6. 1g. Scandium(III) nitrate pentahydrate (99.9%-Sc) (REO). MFCD00149849. Molecular Weight: 230.97 (321.05). Molecular Formula: Sc(NO3)3∙5H2O. Color/form: white xtl. Strem# 93-2104. http://www.strem.com/catalog/v/93-2104/