Learn More
Strem, An Ascensus Company CAS# 12060-08-1. 5g. Scandium(III) oxide (99.9%-Sc) (REO). MFCD00011223
SDP
Catalog No. 5090114407
Encompass
CAS# 12060-08-1. 5g. Scandium(III) oxide (99.9%-Sc) (REO). MFCD00011223. Molecular Weight: 137.91. Molecular Formula: Sc2O3. Color/form: white pwdr. Strem# 93-2113. http://www.strem.com/catalog/v/93-2113/
Catalog No. 50-901-14407 Supplier Strem, An Ascensus Company Supplier No. 9321135G
CAS# 12060-08-1. 5g. Scandium(III) oxide (99.9%-Sc) (REO). MFCD00011223. Molecular Weight: 137.91. Molecular Formula: Sc2O3. Color/form: white pwdr. Strem# 93-2113. http://www.strem.com/catalog/v/93-2113/