Learn More
Strem, An Ascensus Company CAS# 144026-79-9. 1g. Scandium(III) trifluoromethanesulfonate, min. 98% (Scandium triflate). MFCD00192433
SDP
Catalog No. 5090110708
Encompass
CAS# 144026-79-9. 1g. Scandium(III) trifluoromethanesulfonate, min. 98% (Scandium triflate). MFCD00192433. Molecular Weight: 492.17. Molecular Formula: Sc(SO3CF3)3. Color/form: white pwdr. Strem# 21-2000. http://www.strem.com/catalog/v/21-2000/
Catalog No. 50-901-10708 Supplier Strem, An Ascensus Company Supplier No. 2120001G
CAS# 144026-79-9. 1g. Scandium(III) trifluoromethanesulfonate, min. 98% (Scandium triflate). MFCD00192433. Molecular Weight: 492.17. Molecular Formula: Sc(SO3CF3)3. Color/form: white pwdr. Strem# 21-2000. http://www.strem.com/catalog/v/21-2000/