Learn More
Aqua Solutions Scandium Standard 100.0 ppm in 5% v/v HCl, NIST Traceable (1L)
SDP
Catalog No. NC0619831
Encompass
Scandium Standard 100.0 ppm in 5% v/v HCl, NIST Traceable (1L)
Catalog No. NC0619831 Supplier Aqua Solutions Supplier No. MOB090
Scandium Standard 100.0 ppm in 5% v/v HCl, NIST Traceable (1L)