Learn More
Molecular Devices Molecular Devices Assay Kit, Two-Color Fluorescence, Mammalian Cells, 96-Well
Catalog No. 5000014351
Encompass_Preferred
The EarlyTox Live/Dead Assay Kit is a two-color fluorescence assay for simultaneous detection of live and dead mammalian cells based on membrane integrity. Designed for fluorescence microplate readers, the Explorer kit is configured for 96-well workflows.
  • provides two-color fluorescence detection of live and dead cells
  • includes calcein AM and ethidium homodimer-III reagents
  • explorer kit size sufficient for two 96-well microplates
  • recommended plate: black-walled, clear-bottom 96-well microplates
  • excitation/emission: calcein AM 495/530 nm, ethidium homodimer-III 530/645 nm
  • store sealed, desiccated, protected from light at -20°C
  • use aqueous working solutions within one day and test dye concentrations for optimal results
Catalog No. 50-000-14351 Supplier Molecular Devices Supplier No. R8340
The EarlyTox Live/Dead Assay Kit is a two-color fluorescence assay for simultaneous detection of live and dead mammalian cells based on membrane integrity. Designed for fluorescence microplate readers, the Explorer kit is configured for 96-well workflows.
  • provides two-color fluorescence detection of live and dead cells
  • includes calcein AM and ethidium homodimer-III reagents
  • explorer kit size sufficient for two 96-well microplates
  • recommended plate: black-walled, clear-bottom 96-well microplates
  • excitation/emission: calcein AM 495/530 nm, ethidium homodimer-III 530/645 nm
  • store sealed, desiccated, protected from light at -20°C
  • use aqueous working solutions within one day and test dye concentrations for optimal results