Learn More
Molecular Devices Molecular Devices Secondary Antibody, Goat Anti-Rabbit, Eu-Labeled, 60 μg
Catalog No. 5000014461
Encompass_Preferred
The ScanLater Eu-labeled goat anti-rabbit antibody is a europium-chelate-conjugated secondary antibody supplied as a bulk vial for time-resolved fluorescence (TRF) western blot detection. It is formulated for sensitive detection of rabbit primary antibodies in europium-based TRF workflows and is intended for reconstitution and dilution prior to use.
  • Europium-chelate conjugated secondary antibody for TRF detection.
  • Bulk vial containing 60 μg of antibody.
  • Reconstitute bulk vial in 60 μL reagent-grade water.
  • Recommended working dilution for bulk: 1:5,000.
  • Store unmodified vial at -20 °C, protect from light.
  • Aliquot reconstituted material and avoid repeated freeze-thaw cycles.
  • Designed for detection of rabbit primary antibodies in western blot assays.
Catalog No. 50-000-14461 Supplier Molecular Devices Supplier No. R8204
The ScanLater Eu-labeled goat anti-rabbit antibody is a europium-chelate-conjugated secondary antibody supplied as a bulk vial for time-resolved fluorescence (TRF) western blot detection. It is formulated for sensitive detection of rabbit primary antibodies in europium-based TRF workflows and is intended for reconstitution and dilution prior to use.
  • Europium-chelate conjugated secondary antibody for TRF detection.
  • Bulk vial containing 60 μg of antibody.
  • Reconstitute bulk vial in 60 μL reagent-grade water.
  • Recommended working dilution for bulk: 1:5,000.
  • Store unmodified vial at -20 °C, protect from light.
  • Aliquot reconstituted material and avoid repeated freeze-thaw cycles.
  • Designed for detection of rabbit primary antibodies in western blot assays.