Learn More
Molecular Devices Molecular Devices Blocking Buffer, 5X Concentrate, 120 mL, Liquid
Catalog No. 5000014443
Encompass_Preferred
Concentrated 5X blocking buffer formulated for fluorescence western blot detection. Dilute to 1X for membrane blocking prior to primary antibody incubation to reduce background and improve signal-to-noise in fluorescence-based assays.
  • Concentrated 5X formulation.
  • Dilutes to 1X for use as a blocking solution.
  • Supplied as a 120 mL bottle.
  • Reduces background and improves signal-to-noise in fluorescence western blots.
  • Compatible with membrane blocking prior to antibody incubations.
  • For research use only; not for clinical or diagnostic use.
Catalog No. 50-000-14443 Supplier Molecular Devices Supplier No. R8207
Concentrated 5X blocking buffer formulated for fluorescence western blot detection. Dilute to 1X for membrane blocking prior to primary antibody incubation to reduce background and improve signal-to-noise in fluorescence-based assays.
  • Concentrated 5X formulation.
  • Dilutes to 1X for use as a blocking solution.
  • Supplied as a 120 mL bottle.
  • Reduces background and improves signal-to-noise in fluorescence western blots.
  • Compatible with membrane blocking prior to antibody incubations.
  • For research use only; not for clinical or diagnostic use.