Learn More
Molecular Devices Molecular Devices Enhanced TRF Module, Time-Resolved Fluorescence, SpectraMax iD5 Compatible
Catalog No. 5000014728
Encompass_Preferred
The Enhanced TRF Module expands time-resolved fluorescence performance of SpectraMax iD5 and iD5e microplate readers, enabling TRF, TR-FRET, HTRF, IMAP, and ScanLater western blot reads with improved sensitivity for lanthanide assays.
  • Improved sensitivity for Europium detection down to 10 fM.
  • Enables TRF, TR-FRET, and HTRF assay read modes.
  • Supports ScanLater western blot TRF reads.
  • Operates with configurable flash intervals for time-resolved reads.
  • Compatible with exchangeable excitation and emission filter slides sold separately.
  • Requires compatible data acquisition software for some TR-FRET read modes.
Catalog No. 50-000-14728 Supplier Molecular Devices Supplier No. 02007030
Add to Cart
Add to Cart
Request a Quote
The Enhanced TRF Module expands time-resolved fluorescence performance of SpectraMax iD5 and iD5e microplate readers, enabling TRF, TR-FRET, HTRF, IMAP, and ScanLater western blot reads with improved sensitivity for lanthanide assays.
  • Improved sensitivity for Europium detection down to 10 fM.
  • Enables TRF, TR-FRET, and HTRF assay read modes.
  • Supports ScanLater western blot TRF reads.
  • Operates with configurable flash intervals for time-resolved reads.
  • Compatible with exchangeable excitation and emission filter slides sold separately.
  • Requires compatible data acquisition software for some TR-FRET read modes.