Learn More
Molecular Devices Molecular Devices FLIPR Potassium Assay Kit, Thallium Detection, Homogeneous, 96/384-well
Catalog No. 5000014348
Encompass_Preferred
The FLIPR Potassium Assay Kit is a fluorescence-based, cell assay for measuring functional activity of ligand- and voltage-gated potassium channels using a thallium (Tl+) indicator dye. It provides a homogeneous, no-wash protocol with enhanced signal window and improved signal-to-noise, suitable for high-throughput screening on FLIPR systems.
  • Homogeneous, no-wash assay for high-throughput screening.
  • Thallium-based fluorescent detection for functional potassium channel readouts.
  • Designed for ligand-gated and voltage-gated K+ channel measurements.
  • Explorer kit format supplies reagents sufficient for ten 96- or 384-well plates.
  • Includes dye components, assay buffers, K2SO4 and Tl2SO4 solutions.
  • Compatible with FLIPR instrument platforms and adaptable to 1536-well formats.
Catalog No. 50-000-14348 Supplier Molecular Devices Supplier No. R8222
The FLIPR Potassium Assay Kit is a fluorescence-based, cell assay for measuring functional activity of ligand- and voltage-gated potassium channels using a thallium (Tl+) indicator dye. It provides a homogeneous, no-wash protocol with enhanced signal window and improved signal-to-noise, suitable for high-throughput screening on FLIPR systems.
  • Homogeneous, no-wash assay for high-throughput screening.
  • Thallium-based fluorescent detection for functional potassium channel readouts.
  • Designed for ligand-gated and voltage-gated K+ channel measurements.
  • Explorer kit format supplies reagents sufficient for ten 96- or 384-well plates.
  • Includes dye components, assay buffers, K2SO4 and Tl2SO4 solutions.
  • Compatible with FLIPR instrument platforms and adaptable to 1536-well formats.