Learn More
Motic x ISENET Scanner TMA Advanced Research ImF Package with EasyScan FlexScan60 and Galileo TMAtic
Shop All Motic Group ProductsDescription
- Motic EasyScan FlexScan60 digital slide scanner for research imaging workflows.
- Galileo TMAtic fully automated TMA construction platform.
- Automated needle cleaning and integrity checks to support consistent TMA construction.
- Digital overlay for tissue-region selection and core-position documentation.
- Workflow designed to support larger TMA projects and downstream digital image analysis.
- Combines automated tissue array construction with digital imaging for advanced research.
- Helps improve workflow consistency across large sample sets.
- Supports standardized comparison of tissue markers and experimental conditions.
- Facilitates the organization and review of image data from multiple tissue samples.
- Provides a foundation for research workflows involving fluorescence imaging and quantitative analysis, subject to the scanner configuration.
- Immunofluorescence (IF) research, subject to supported scanner configuration.
- Multiplex fluorescence and biomarker-expression studies.
- Cancer biology and tumor microenvironment research.
- Comparative tissue analysis across large cohorts.
- Advanced pathology research and digital image quantification.
Specifications
Specifications
| Includes | Fully Automated Platform |
| Product Type | Fluorescence-based Digital Scanner |
| For Use With (Equipment) | Motic EasyScan FlexScan60 and Galileo TMAtic |