Learn More
Motic x ISENET Scanner TMA Basic Package with EasyScan 2 and Galileo TMA CK2500
Shop All Motic Group ProductsDescription
- Motic EasyScan Two digital slide scanning system.
- Galileo CK2500 semi-automatic TMA construction system.
- Touchscreen-controlled stage for operator-guided TMA construction.
- Capacity for up to six standard tissue cassettes, with up to five replicas.
- Interchangeable punch needle options for different tissue-core sizes.
- Provides an accessible starting point for laboratories adopting digital pathology and TMA techniques.
- Simplifies operator-guided tissue array construction.
- Supports routine sample preparation and repeatable TMA layouts.
- Offers a practical workflow for smaller research projects and training environments.
- Helps laboratories digitize tissue slides for convenient viewing, sharing, and analysis.
- Routine histology and tissue morphology research.
- Introductory TMA research and laboratory training.
- Basic biomarker screening and validation.
- IHC and comparative staining studies.
- Small-cohort pathology research and digital slide review.
Specifications
Specifications
| Includes | Semi-automatic Platform |
| Product Type | Digital Scanner |
| For Use With (Equipment) | Motic EasyScan Two and Galileo TMA CK2500 |