Learn More
Motic x ISENET Scanner TMA Pharma Package with EasyScan Pro 24 and Galileo TMA CK4600
Shop All Motic Group ProductsDescription
- Motic EasyScan Pro 24 for mid-volume digital slide scanning.
- Galileo CK4600 computer-driven TMA construction system.
- Multiple tissue cassette configurations to accommodate different sample requirements.
- Flexible core selection using tissue images, manual overlays, or digital overlays.
- Digital reporting of donor-block images and extracted core positions.
- Supports flexible TMA designs for complex research studies.
- Helps standardize tissue sampling across treatment groups, controls, and experimental cohorts.
- Facilitates the documentation and traceability of tissue cores.
- Enables efficient digital review of prepared tissue slides.
- Supports research workflows requiring reproducible sample selection and comparative analysis.
- Pharmaceutical and preclinical drug research.
- Drug-response and treatment-group comparison studies.
- Biomarker validation and target-expression research.
- IHC and multiplex staining studies.
- Tumor biology and translational research.
Specifications
Specifications
| Includes | Computer-driven Platform |
| Product Type | Digital Scanner |
| For Use With (Equipment) | Motic EasyScan Pro 24 and Galileo TMA CK4600 |