Learn More
Motic x ISENET Scanner TMA Research ImF Package with EasyScan FlexScan60 and Galileo TMA CK4600
Shop All Motic Group ProductsDescription
- Motic EasyScan FlexScan60 digital slide scanner for research imaging workflows.
- Galileo CK4600 computer-driven TMA system for flexible array design.
- Multiple core sizes and flexible tissue cassette configurations.
- Digital tissue-region selection and core-position reporting.
- Supports customized TMA layouts for downstream research and image analysis.
- Provides greater flexibility for specialized TMA research designs.
- Supports precise tissue sampling for comparative experimental studies.
- Helps maintain traceability between donor tissues and constructed arrays.
- Facilitates digital review of tissue samples and research findings.
- Suitable for laboratories requiring adaptable TMA construction rather than fully automated, high-throughput processing.
- Immunofluorescence (IF) research, subject to supported scanner configuration.
- Multiplex biomarker and protein-expression studies.
- Cancer research and tumor heterogeneity studies.
- Preclinical research and comparative tissue analysis.
- Digital pathology and quantitative image analysis.
Specifications
Specifications
| Includes | Computer-driven Platform |
| Product Type | Fluorescence-based Digital Scanner |
| For Use With (Equipment) | Motic EasyScan FlexScan60 and Galileo TMA CK4600 |