Learn More
Motic x ISENET Scanner TMA Routine Package with EasyScan Pro 24 and Galileo TMA CK3600
Shop All Motic Group ProductsDescription
- Motic EasyScan Pro 24 digital slide scanner for efficient whole-slide imaging.
- Galileo TMA CK3600 system for tissue microarray (TMA) construction.
- Integrated workflow combining tissue array preparation and digital slide scanning.
- Supports organized tissue sampling and preparation for downstream digital analysis.
- Designed to support routine laboratory research and tissue analysis workflows.
- Streamlines tissue microarray preparation and digital slide imaging.
- Helps standardize tissue sampling for comparative research studies.
- Reduces repetitive manual work in routine TMA workflows.
- Facilitates digital slide review, image sharing, and sample documentation.
- Supports efficient analysis of multiple tissue samples in research laboratories.
- Routine histology and tissue morphology research.
- Tissue microarray (TMA) preparation and analysis.
- Immunohistochemistry (IHC) and biomarker screening.
- Comparative tissue studies and research cohort analysis.
- Cancer biology and translational pathology research.
- Digital pathology and quantitative image analysis.
Specifications
Specifications
| Includes | Computer-assisted Platform |
| Product Type | Digital Scanner |
| For Use With (Equipment) | Motic EasyScan Pro 24 and Galileo TMA CK3600 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.