Learn More
Fine Science Tools Micro Knives - Plastic Handle, 15° Cutting Angle, 13 cm
SDP
Catalog No. NC9786299
Encompass
Product: Micro Knife - Plastic Handle/15 deg. Cutting Angle/13cm/ Blade Shape: 15 deg. Cutting Angle/ Cutting Edge: 5mm/ Thickness: 0.15mm/ Alloy / Material: Carbon Steel Tip / Plastic Handle
Catalog No. NC9786299 Supplier Fine Science Tools Supplier No. 1031512
Product: Micro Knife - Plastic Handle/15 deg. Cutting Angle/13cm/ Blade Shape: 15 deg. Cutting Angle/ Cutting Edge: 5mm/ Thickness: 0.15mm/ Alloy / Material: Carbon Steel Tip / Plastic Handle