Learn More
Aviva Systems Biology Corp SCAPER Antibody - N-terminal region (ARP39448_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39448_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCAPER. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SCAPER. Alternate names: FLJ31533, FLJ31953, FLJ46212, KIAA1454, MSTP063, ZNF291, Zfp291ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-807