Learn More
Alkali Scientific ASI™ Scar™ Block 35mm Cell Culture Dish, PS, Sterile, 30/Pk, 2 Well
SDP
Catalog No. 503026477
Encompass_Preferred
ASIScar™ Block is easily attachable to surface, and thus applicable to a variety of products for studying cell migration, wound healing, and metastasis of cancer cell. Block is composed of 500㎛ thick walls to artificially generate cell free gap(s), insuring higher uniformity and reproducibility. Packed individually Non-pyrogenic Non-cytotoxic DNase / RNase-free Human DNA-free
Catalog No. 50-302-6477 Supplier Alkali Scientific Supplier No. CCD002
ASIScar™ Block is easily attachable to surface, and thus applicable to a variety of products for studying cell migration, wound healing, and metastasis of cancer cell. Block is composed of 500㎛ thick walls to artificially generate cell free gap(s), insuring higher uniformity and reproducibility. Packed individually Non-pyrogenic Non-cytotoxic DNase / RNase-free Human DNA-free