Learn More
Alkali Scientific ASI™ Scar™ Block 4 Well Cell Culture Slide, Sterile, 24/Pk, Glass
SDP
Catalog No. 503026609
Encompass_Preferred
ASIScar™ Block is easily attachable to surface, and thus applicable to a variety of products for studying cell migration, wound healing, and metastasis of cancer cell. Block is composed of 500㎛ thick walls to artificially generate cell free gap(s), insuring higher uniformity and reproducibility. Packed individually Non-pyrogenic Non-cytotoxic DNase / RNase-free Human DNA-free
Catalog No. 50-302-6609 Supplier Alkali Scientific Supplier No. CCS04G
ASIScar™ Block is easily attachable to surface, and thus applicable to a variety of products for studying cell migration, wound healing, and metastasis of cancer cell. Block is composed of 500㎛ thick walls to artificially generate cell free gap(s), insuring higher uniformity and reproducibility. Packed individually Non-pyrogenic Non-cytotoxic DNase / RNase-free Human DNA-free