Learn More
Alkali Scientific ASI™ Scar Scratcher, 6 Well, 1.0mm
SDP
Catalog No. 503026930
Encompass_Preferred
Alkali's Scar Scratcher improves the uniformity and reproducibility of the possible products used in cancer metastasis, wound healing and cell migration research. Alkali's Scar Scratcher consists of an alignment lid (individually wrapped / STERILE) with identical holes and a scratcher (individually wrapped / STERILE), suitable for use in both 6 well and 24 well configurations. Packing: 20/CASE  Non – pyrogenic Non – cytotoxic DNase / RNase – free Human DNA – free    
Catalog No. 50-302-6930 Supplier Alkali Scientific Supplier No. SCAR7
Alkali's Scar Scratcher improves the uniformity and reproducibility of the possible products used in cancer metastasis, wound healing and cell migration research. Alkali's Scar Scratcher consists of an alignment lid (individually wrapped / STERILE) with identical holes and a scratcher (individually wrapped / STERILE), suitable for use in both 6 well and 24 well configurations. Packing: 20/CASE  Non – pyrogenic Non – cytotoxic DNase / RNase – free Human DNA – free    