Learn More
Alkali Scientific Scar Scratcher with 24-Well Lid, HDPE, 0.5mm, Individually Wrapped, Sterile, 20/cs
SDP
Catalog No. 502389242
Encompass_Preferred
Scar Scratcher improves the uniformity and reproducibility of the possible products used in cancer metastasis, wound healing and cell migration research. Scar Scratcher consists of a lid with identical holes and a scratcher, suitable for use in 24-well configurations.
Catalog No. 50-238-9242 Supplier Alkali Scientific Supplier No. SCAR24
Scar Scratcher improves the uniformity and reproducibility of the possible products used in cancer metastasis, wound healing and cell migration research. Scar Scratcher consists of a lid with identical holes and a scratcher, suitable for use in 24-well configurations.