Learn More
Alkali Scientific Scar Scratcher with 48-Well Lid, HDPE, 1mm, Individually Wrapped, Sterile, 20/pk
SDP
Catalog No. 502388774
Encompass_Preferred
Scar Scratcher improves the uniformity and reproducibility of the possible products used in cancer metastasis, wound healing and cell migration research. Scar Scratcher consists of a lid with identical holes and a scratcher, suitable for use in 48-well configurations.
Catalog No. 50-238-8774 Supplier Alkali Scientific Supplier No. SCAR26
Scar Scratcher improves the uniformity and reproducibility of the possible products used in cancer metastasis, wound healing and cell migration research. Scar Scratcher consists of a lid with identical holes and a scratcher, suitable for use in 48-well configurations.