Learn More
Alkali Scientific Scar Scratcher with 6-Well Lid, HDPE, 0.5mm, Individually Wrapped, Sterile, 20/pk
SDP
Catalog No. 502389000
Encompass_Preferred
Scar Scratcher improves the uniformity and reproducibility of the possible products used in cancer metastasis, wound healing and cell migration research. Scar Scratcher consists of a lid with identical holes and a scratcher, suitable for use in 6-well configurations.
Catalog No. 50-238-9000 Supplier Alkali Scientific Supplier No. SCAR6
Scar Scratcher improves the uniformity and reproducibility of the possible products used in cancer metastasis, wound healing and cell migration research. Scar Scratcher consists of a lid with identical holes and a scratcher, suitable for use in 6-well configurations.