Learn More
Aviva Systems Biology Corp Scara3 Antibody - middle region (ARP47233_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47233_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Scara3. It was validated on Western Blot. . Alternate names: APC7, C130058N24Rik, CSR, CSR1, MSLR1, MSRL1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-291