Learn More
ACROBiosystems Recombinant Protein;Human SCARB1 / SR-B1 Protein, Fc Tag;HEK293;50UG;SC1,SCARB1,CD36L1,SRB1,CLA-1,SR-BI,CLA1,HDLQTL6,SCARB-1,SRB-1,SRBI,CD36 L1
Supplier:  ACROBiosystems SC1H525850UG
Encompass_Preferred
Recombinant Protein;Human SCARB1 / SR-B1 Protein, Fc Tag;50UG;Human SCARB1, Fc Tag (SC1-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 33 - Tyr 443 (Accession # NP_005496.4).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.6 kDa. As a result of glycosylation, the protein migrates as 110-115 kDa under reducing (R) condition, and 220-230 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8426