Learn More
Aviva Systems Biology Corp SCARB2 Antibody - N-terminal region (ARP61682_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61682_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCARB2. It was validated on Western Blot. . Alternate names: AMRF, CD36L2, HLGP85, LIMPII, SR-BII, EPM4, LGP85, LIMP-2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-438