Learn More
ACROBiosystems Human SCARB2 Protein, Fc Tag, Lyophilized, 1 mg
Catalog No. 501658498
Encompass_Preferred
Human SCARB2, also known as LIMP2, is a member of the CD36 family that functions as a lysosomal receptor for glucosylceramidase (GBA) targeting. This protein is expressed in human HEK293 cells and contains amino acids Arg 27 to Thr 432, with a human IgG1 Fc tag at the C-terminus.
  • Purity >85% as determined by SDS-PAGE
  • Calculated molecular weight of 72.6 kDa, migrating as 90–115 kDa under reducing conditions due to glycosylation
  • Endotoxin levels <1.0 EU per μg
  • Lyophilized in 50 mM Tris, 100 mM glycine, 25 mM arginine, 150 mM NaCl, pH 7.5 with trehalose
  • Stable for 12 months at -20°C to -70°C in lyophilized state
  • Please refer to the Certificate of Analysis for specific reconstitution instructions
Catalog No. 50-165-8498 Supplier ACROBiosystems Supplier No. SC2H52501MG
Human SCARB2, also known as LIMP2, is a member of the CD36 family that functions as a lysosomal receptor for glucosylceramidase (GBA) targeting. This protein is expressed in human HEK293 cells and contains amino acids Arg 27 to Thr 432, with a human IgG1 Fc tag at the C-terminus.
  • Purity >85% as determined by SDS-PAGE
  • Calculated molecular weight of 72.6 kDa, migrating as 90–115 kDa under reducing conditions due to glycosylation
  • Endotoxin levels <1.0 EU per μg
  • Lyophilized in 50 mM Tris, 100 mM glycine, 25 mM arginine, 150 mM NaCl, pH 7.5 with trehalose
  • Stable for 12 months at -20°C to -70°C in lyophilized state
  • Please refer to the Certificate of Analysis for specific reconstitution instructions