Learn More
Biotium SCAS
Catalog No. NC1669436
Encompass_Preferred
SCAS (4-Sulfonato calix[8]arene, sodium salt) is a quencher developed by Biotium to reduce background fluorescence when using our fixable AM dyes, SynaptoGreen and SynaptoRed dyes. Unlike ADVASEP-7, SCAS dramatically lowers background fluorescence as soon as it is added to the preparation without the need for repeated wash steps. Properties: Off-white solid soluble in water; C56H40Na8O32S8; MW 1665.34
Catalog No. NC1669436 Supplier Biotium Supplier No. 70037
SCAS (4-Sulfonato calix[8]arene, sodium salt) is a quencher developed by Biotium to reduce background fluorescence when using our fixable AM dyes, SynaptoGreen and SynaptoRed dyes. Unlike ADVASEP-7, SCAS dramatically lowers background fluorescence as soon as it is added to the preparation without the need for repeated wash steps. Properties: Off-white solid soluble in water; C56H40Na8O32S8; MW 1665.34