Learn More
Haematologic Technologies Inc CTI vacuum collection tube | 5mL
Catalog No. NC1756746
Encompass
This supplier has a minimum order requirement of 25 each for this item. To simplify the process of collecting samples with added CTI, HTI has developed blood collection tubes containing CTI. These tubes simplify the process of conducting TF-dependent studies by allowing you to draw blood directly onto an anticoagulant containing CTI
Catalog No. NC1756746 Supplier Haematologic Technologies Inc Supplier No. SCAT27 4.5/5
This supplier has a minimum order requirement of 25 each for this item. To simplify the process of collecting samples with added CTI, HTI has developed blood collection tubes containing CTI. These tubes simplify the process of conducting TF-dependent studies by allowing you to draw blood directly onto an anticoagulant containing CTI