Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLINGSHOT BIOSCIENCES ScatterBridge, Hydrogel, 9 populations (3×3 grid), 25 tests
SDP

Catalog No. NC2742616
Encompass_Preferred

ScatterBridge Scatter Controls are a synthetic hydrogel reference control with nine defined scatter populations designed to standardize forward and side scatter, support linearity assessments and track instrument performance over time.

Specifications:
Catalog Number: SL-00027
Format: Hydrogel, 9 populations (3×3 grid)
Package Size: 25 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: Wet Ice
Shelf Life: 12 months
Country of Origin: United States
For Research Use Only.

Catalog No. NC2742616 Supplier SLINGSHOT BIOSCIENCES Supplier No. SL00027
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ScatterBridge Scatter Controls are a synthetic hydrogel reference control with nine defined scatter populations designed to standardize forward and side scatter, support linearity assessments and track instrument performance over time.

Specifications:
Catalog Number: SL-00027
Format: Hydrogel, 9 populations (3×3 grid)
Package Size: 25 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: Wet Ice
Shelf Life: 12 months
Country of Origin: United States
For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.