Learn More
Medchemexpress LLC Scavenger receptor class B member 1 (human, recombinant, HEK293, His-Fc) | 95.0% | 5 UG
SDP
Catalog No. 5000290499
Encompass_Preferred
Recombinant human scavenger receptor class B member 1 (SR-BI/SCARB1) extracellular domain produced in HEK293 cells and supplied as a glycosylated, His-Fc-tagged protein. Supplied as a purified preparation for biochemical assays, receptor-ligand interaction studies, and antibody validation.
  • Expressed in HEK293 cells.
  • C-terminal His-Fc tag for detection and capture.
  • Glycosylated protein with approximate molecular weight 100-120 kDa.
  • Purity approximately 95.0%.
  • Suitable for biochemical assays, binding studies, and antibody characterization.
  • Available in small research pack sizes, for example 5 μg.
Catalog No. 50-002-90499 Supplier Medchemexpress LLC Supplier No. HYP766585UG
Recombinant human scavenger receptor class B member 1 (SR-BI/SCARB1) extracellular domain produced in HEK293 cells and supplied as a glycosylated, His-Fc-tagged protein. Supplied as a purified preparation for biochemical assays, receptor-ligand interaction studies, and antibody validation.
  • Expressed in HEK293 cells.
  • C-terminal His-Fc tag for detection and capture.
  • Glycosylated protein with approximate molecular weight 100-120 kDa.
  • Purity approximately 95.0%.
  • Suitable for biochemical assays, binding studies, and antibody characterization.
  • Available in small research pack sizes, for example 5 μg.