Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Scavenger receptor class B member 1 (SR-BI/SCARB1), recombinant human | >95.0% | 10 UG
SDP

Catalog No. 5000290504
Encompass_Preferred

Recombinant human scavenger receptor class B member 1 (SR-BI/SCARB1) expressed in HEK293 cells and supplied as a lyophilized C-terminal His-tagged protein. Supplied as a 10 μg lyophilized vial, the product runs at ~70-90 kDa and is >95% pure, suitable for biochemical and cell biology assays requiring recombinant human SR-BI.

  • Human sequence P33-Y443, accession Q8WTV0-1.
  • C-terminal His tag facilitates purification and detection.
  • Expressed in HEK293 for mammalian post-translational modifications.
  • Lyophilized with stabilizers (trehalose or mannitol, and Tween 80) for stability.
  • Purity >95.0% by reducing SDS-PAGE for consistent performance.
  • Reconstitute to ≥100 μg/mL; avoid concentrations below 100 μg/mL in ddH2O.

Catalog No. 50-002-90504 Supplier Medchemexpress LLC Supplier No. HYP7665810UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human scavenger receptor class B member 1 (SR-BI/SCARB1) expressed in HEK293 cells and supplied as a lyophilized C-terminal His-tagged protein. Supplied as a 10 μg lyophilized vial, the product runs at ~70-90 kDa and is >95% pure, suitable for biochemical and cell biology assays requiring recombinant human SR-BI.

  • Human sequence P33-Y443, accession Q8WTV0-1.
  • C-terminal His tag facilitates purification and detection.
  • Expressed in HEK293 for mammalian post-translational modifications.
  • Lyophilized with stabilizers (trehalose or mannitol, and Tween 80) for stability.
  • Purity >95.0% by reducing SDS-PAGE for consistent performance.
  • Reconstitute to ≥100 μg/mL; avoid concentrations below 100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.