Learn More
Cayman Chemical Scavenger Receptor B2/CD36 Blocking Peptide, 1 ea
SDP
Catalog No. 502988707
Encompass_Preferred
Peptide Sequence: Human CD36 sequence amino acids 98-114 (AKENVTQDAEDNTVSF) • To be used in conjunction with Cayman’s CD36 polyclonal antibody (Item No. 100011) to block protein-antibody complex formation during immunochemical analysis of CD36.
Catalog No. 50-298-8707 Supplier Cayman Chemical Supplier No. 3000111 EA
Peptide Sequence: Human CD36 sequence amino acids 98-114 (AKENVTQDAEDNTVSF) • To be used in conjunction with Cayman’s CD36 polyclonal antibody (Item No. 100011) to block protein-antibody complex formation during immunochemical analysis of CD36.