Learn More
Cayman Chemical Scavenger Receptor B2/CD36 Extracellular Domain (mouse, recombinant), Purity: ≥92% estimated by SDS-PAGE, 1 mg
SDP
Catalog No. 502910205
Encompass_Preferred
Source: Active recombinant mouse C-terminal His-tagged CD36 expressed in HEK293 cells • Amino acids: 30-439 • MW: 47.8 kDa
Catalog No. 50-291-0205 Supplier Cayman Chemical Supplier No. 357291 MG
Source: Active recombinant mouse C-terminal His-tagged CD36 expressed in HEK293 cells • Amino acids: 30-439 • MW: 47.8 kDa