Learn More
Cayman Chemical Scavenger Receptor B2/CD36 Monoclonal Antibody (Clone JC63.1), 1 ea
SDP
Catalog No. 502966186
Encompass_Preferred
Immunogen: Recombinant full-length mouse CD36 expressed in adenovirus • Host: CD36 null mouse • Species Reactivity: (+) Human, mouse, rat • Cross Reactivity: (+) CD36 • Applications: FC
Catalog No. 50-296-6186 Supplier Cayman Chemical Supplier No. 1881501 EA
Immunogen: Recombinant full-length mouse CD36 expressed in adenovirus • Host: CD36 null mouse • Species Reactivity: (+) Human, mouse, rat • Cross Reactivity: (+) CD36 • Applications: FC