Learn More
Cayman Chemical ScavengR RECPTR B2CD36 Ex
SDP
Catalog No. 502911399
Encompass_Preferred
Source: Active recombinant mouse C-terminal His-tagged CD36 expressed in HEK293 cells • Amino acids: 30-439 • MW: 47.8 kDa
Catalog No. 50-291-1399 Supplier Cayman Chemical Supplier No. 35729100 UG
Source: Active recombinant mouse C-terminal His-tagged CD36 expressed in HEK293 cells • Amino acids: 30-439 • MW: 47.8 kDa