Learn More
Cayman Chemical ScavengR RECPTR B2CD36 Ig
SDP
Catalog No. 502948596
Encompass_Preferred
Preparation: This antibody, which contains a mouse IgG1 isoform, was produced as a recombinant protein expressed in HEK293 cells using an engineered construct based on a monoclonal scavenger receptor B2/CD36 antibody • Host: HEK293 cells • Species Reactivity: (+) Human, mouse, rat • Cross Reactivity: (+) CD36 • Applications: FC
Catalog No. 50-294-8596 Supplier Cayman Chemical Supplier No. 33938100 UG
Preparation: This antibody, which contains a mouse IgG1 isoform, was produced as a recombinant protein expressed in HEK293 cells using an engineered construct based on a monoclonal scavenger receptor B2/CD36 antibody • Host: HEK293 cells • Species Reactivity: (+) Human, mouse, rat • Cross Reactivity: (+) CD36 • Applications: FC